Learn More
Aviva Systems Biology Corp SOX13 Antibody - C-terminal region (ARP33728_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33728_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SOX13. Alternate names: ICA12, Sox-13ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55125