Learn More
Aviva Systems Biology Corp SOX14 Antibody - N-terminal region (ARP35711_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35711_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SOX14. Alternate names: SOX28ApplicationWB. Species reactivity: Cow, Goat, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55825