Learn More
Aviva Systems Biology Corp SOX15 Antibody - middle region (ARP38883_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38883_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX15. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SOX15. Alternate names: SOX20, SOX26, SOX27ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56832