Learn More
Aviva Systems Biology Corp SOX15 Antibody - N-terminal region (ARP38882_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38882_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SOX15. Alternate names: SOX20, SOX26, SOX27ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-014-56831