Learn More
Aviva Systems Biology Corp SOX15 Antibody - middle region (ARP33329_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33329_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SOX15. Alternate names: SOX20, SOX26, SOX27ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54963