Learn More
Aviva Systems Biology Corp SOX17 Antibody - C-terminal region (ARP33271_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33271_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SOX17. Alternate names: VUR3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-54938