Learn More
Aviva Systems Biology Corp SOX18 Antibody - middle region (ARP33057_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33057_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SOX18. Alternate names: HLTSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54865