Learn More
Aviva Systems Biology Corp SOX18 Antibody - middle region (ARP33056_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33056_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX18. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SOX18. Alternate names: HLTSApplicationIF, WB, IHC. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54864