Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6657MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: SOX2/1792. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: MCF-7 cells or Cervical carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human SOX2 protein fragment (aa176-305). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 34kDa.Chromosome: 3q26.33. Gene Symbol: SOX2. Gene ID: 6657. Ensemble: ENSG00000181449. Uniprot: NP_003097.
Catalog No. 50-188-5180