Learn More
Aviva Systems Biology Corp SOX2 Antibody - middle region (ARP38233_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38233_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SOX2. Alternate names: ANOP3, MCOPS3, MGC2413ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56574