Learn More
Biomatik Corporation SOX2 Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50335
Encompass_Preferred
SOX2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Sry (Sex Determining Region Y)-Box 2. The SOX2 Polyclonal Antibody has been validated for the following applications: ELISA, IF, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Please Inquire. Applications: ELISA, IF, WB, IHC. Target Synonyms: Sry (Sex Determining Region Y)-Box 2. Accession Number: P48431. Gene ID: 6657. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6589