Learn More
Biomatik Corporation Sox2 Recombinant Antibody
Supplier:  Biomatik Corporation RAC07066
Encompass_Preferred
Sox2 Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to SOX2. The Sox2 Recombinant Antibody has been validated for the following applications: ELISA, WB, IHC. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide Derived From Human Sox2. Applications: ELISA, WB, IHC. Target Synonyms: SOX2. Accession Number: P48431. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6587