Learn More
Aviva Systems Biology Corp SOX21 Antibody - middle region (ARP31519_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31519_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SOX21. Alternate names: SOX25ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Zebrafish
Catalog No. 50-014-54253