Learn More
Cayman Chemical Sox2CTerm RabbIt Monclon 100uL
SDP
Catalog No. 502913761
Encompass_Preferred
A flavonoid glycoside with diverse biological activities inhibits CAVII (Ki 4 6 nM) scavenges DPPH and superoxide radicals (IC50s 12 8 and 42 UM respectively) cytotoxic to Jurkat HepG2 and COLO 205 cells (IC50s 11 6 14 3 and 55 4 UM respectively) reduces TPA-induced ear edema (ID50 0 357 mg/ear) decreases triglyceride accumulation in the liver and skeletal muscle in high-fat diet-fed KKAy diabetic mice at 100 mg/kg
Catalog No. 50-291-3761 Supplier Cayman Chemical Supplier No. 35319100 UL
A flavonoid glycoside with diverse biological activities inhibits CAVII (Ki 4 6 nM) scavenges DPPH and superoxide radicals (IC50s 12 8 and 42 UM respectively) cytotoxic to Jurkat HepG2 and COLO 205 cells (IC50s 11 6 14 3 and 55 4 UM respectively) reduces TPA-induced ear edema (ID50 0 357 mg/ear) decreases triglyceride accumulation in the liver and skeletal muscle in high-fat diet-fed KKAy diabetic mice at 100 mg/kg