Learn More
Aviva Systems Biology Corp SOX3 Antibody - C-terminal region (ARP38649_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38649_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SOX3. Alternate names: PHP, GHDX, MRGH, PHPX, SOXBApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56739