Learn More
Aviva Systems Biology Corp SOX3 Antibody - N-terminal region (ARP33321_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33321_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SOX3. Alternate names: PHP, GHDX, MRGH, PHPX, SOXBApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-54959