Learn More
Aviva Systems Biology Corp SOX30 Antibody - middle region (ARP32990_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32990_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX30. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SOX30. Alternate names: -ApplicationWB. Species reactivity: Dog, Human, Pig
Catalog No. 50-014-54845