Learn More
Aviva Systems Biology Corp SOX4 Antibody - N-terminal region (ARP33252_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33252_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SOX4. Alternate names: EVI16ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit
Catalog No. 50-014-54933