Learn More
Aviva Systems Biology Corp SOX6 Antibody - middle region (ARP33148_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33148_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SOX6. Alternate names: HSSOX6, SOXDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54896