Learn More
Aviva Systems Biology Corp SOX7 Antibody - middle region (ARP39668_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39668_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SOX7. Alternate names: MGC10895ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57130