Learn More
Aviva Systems Biology Corp SOX9 Antibody - N-terminal region (P100797_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100797_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SOX9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SOX9. Alternate names: CMD1, SRA1, CMPD1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62716