Learn More
Abcam Anti-SOX9 antibody [EPR14335-78], 100 μL, Rabbit monoclonal, Purified, IgG, Unconjugated, Reactivity: Human, Mouse, Rat
Supplier:  Abcam AB185966100UL
Encompass_Preferred
Anti-SOX9 antibody [EPR14335-78], 100 μL, Rabbit monoclonal, Purified, IgG, Unconjugated, Reactivity: Human, Mouse, Rat
Catalog No. NC2008099