Learn More
Biotium SOX9 / SRY-box 9(rSOX9/2288), Biotin conjugate, 0.1mg/mL
Supplier:  Biotium BNCB2288100
Encompass_Preferred
The specificity of this monoclonal antibody to its intended target was validated by HuProt™ Array, containing more than 19, 000, full-length human proteins. Plays an important role in the normal skeletal development. May regulate the expression of other genes involved in chondrogenesis by acting as a transcription factor for these genes. Nucleus (Potential). Campomelic dysplasia (CMD1) : Rare, often lethal, dominantly inherited, congenital osteo-chondrodysplasia, associated with male-to-female autosomal sex reversal in two-thirds of the affected karyotypic males. A disease of the newborn characterized by congenital bowing and angulation of long bones, unusually small scapulae, deformed pelvis and spine and a missing pair of ribs. Craniofacial defects such as cleft palate, micrognathia, flat face and hypertelorism are common. Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and phot
Catalog No. 50-006-89053