Learn More
Chemglass Life Sciences Small Extraction Apparatus Complete, Soxhlet, 34/45, 24/40
SDP
Catalog No. CG136901
Encompass_Preferred
Extraction Apparatus, Soxhlet, Small, 34/45, 24/40. Similar to CG-1368, but supplied with a round bottom instead of a flat bottom flask. All specifications are the same as CG-1368.
Catalog No. CG136901 Supplier Chemglass Life Sciences Supplier No. CG136901
Extraction Apparatus, Soxhlet, Small, 34/45, 24/40. Similar to CG-1368, but supplied with a round bottom instead of a flat bottom flask. All specifications are the same as CG-1368.