Learn More
DWK Life Sciences Kimble™ Soxhlet Extraction Body
SDP
Catalog No. K5860000021 Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
Soxhlet Extractor Body
Catalog No. Product Type
K5860000021 Soxhlet Extractor Body
1 options
Catalog No. K5860000021 Supplier DWK Life Sciences Supplier No. 5860000021
Add to Cart
Add to Cart
Extractor body only, used as a component part of 585050 Soxhlet extraction assemblies.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Product Type Soxhlet Extractor Body