Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma Soy Bean Meal (N, S)

Catalog No. AR201630G Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
30 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AR201630G 30 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AR201630G Supplier MilliporeSigma Supplier No. AR201630G
Add to Cart
Add to Cart

Soy Bean Meal (N, S) is a reference standard produced in accordance to ISO Guide 34-2009.

  • Soy Bean Meal (N, S) is a reference standard produced in accordance to ISO Guide 34-2009.
  • This reference has an indefinite known shelf life unopened.
  • Once opened the shelf life is limited to two years.
  • Keep sealed and store under normal dry laboratory conditions.
  • This is a certified reference material (Working Standard) and is traceable to the above-mentioned reference standard.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Soy Bean Meal (N, S)
Type Analytical Standard
Industry Type Food and Beverage
Quantity 30 g
Grade Analytical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.