Learn More
Avanti Polar Lipids Soy Lyso PC | 97281-38-4 | MFCD00131444 | 25mg
Catalog No. NC1616505
Encompass
Soy Lyso PC | Putity: >99% LPC; may contain up to 10% of the 2-LPC isomer | MW: 513.564 | 97281-38-4 | MFCD00131444 | 25mg
Catalog No. NC1616505 Supplier Avanti Polar Lipids Supplier No. 840072P25MG
Soy Lyso PC | Putity: >99% LPC; may contain up to 10% of the 2-LPC isomer | MW: 513.564 | 97281-38-4 | MFCD00131444 | 25mg