Learn More
Bioworld Soy Peptone Yeast Extract Agar, 500 g
Catalog No. 50254479
Encompass_Preferred
Soy Peptone Yeast Extract AgarAgar medium used for the isolation and cultivation of dermatophytes and other pathogenic fungi from clinical specimenspH: 6.6Dehydrated Appearance: homogeneous powderPrepared Appearance: light to dark, yellow to tan agarSoluble in boiling waterContains:Papaic Digest of Soybean Meal Yeast Extract Dextrose Streptomycin Chloramphenicol Agar
Catalog No. 50-254-479 Supplier Bioworld Supplier No. 306260071
Soy Peptone Yeast Extract AgarAgar medium used for the isolation and cultivation of dermatophytes and other pathogenic fungi from clinical specimenspH: 6.6Dehydrated Appearance: homogeneous powderPrepared Appearance: light to dark, yellow to tan agarSoluble in boiling waterContains:Papaic Digest of Soybean Meal Yeast Extract Dextrose Streptomycin Chloramphenicol Agar