Learn More
MSC Zep Degreaser, Soy Response, Heavy-Duty, Liquid, 5 gal
Catalog No. 502817475
Encompass
Soy Response liquid is a powerful, biodegradable degreaser formulated with surfactants, emulsifiers, and a soy-based solvent. Designed for industrial and commercial cleaning, it effectively tackles heavy-duty degreasing jobs on floors, equipment, drains, and grease traps.
  • Powerful liquid degreaser for industrial and commercial use
  • Biodegradable formulation with soy-based solvent
  • High flash point of over 200°F
  • Low VOC formulation meets state and federal regulations
  • Rinses easily with water, leaving no residue
  • Suitable for a variety of surfaces and applications
Catalog No. 50-281-7475 Supplier MSC Supplier No. 11395092
Soy Response liquid is a powerful, biodegradable degreaser formulated with surfactants, emulsifiers, and a soy-based solvent. Designed for industrial and commercial cleaning, it effectively tackles heavy-duty degreasing jobs on floors, equipment, drains, and grease traps.
  • Powerful liquid degreaser for industrial and commercial use
  • Biodegradable formulation with soy-based solvent
  • High flash point of over 200°F
  • Low VOC formulation meets state and federal regulations
  • Rinses easily with water, leaving no residue
  • Suitable for a variety of surfaces and applications