Learn More
Worthington Biochemical Corporation Trypsin Inhibitor, Soybean, Purified, 100mg
SDP
Catalog No. NC9323552
Encompass
Source: Soybean. Animal Free/AF. Chromatographically purified as described by Frattali, V., and Steiner, R.: Biochem., 7, 521 (1968), modified by Worthington. A dialyzed, lyophilized powder. Purity >90% checked using SDS PAGE. Store at 2-8°C.
Catalog No. NC9323552 Supplier Worthington Biochemical Corporation Supplier No. LS003570
Source: Soybean. Animal Free/AF. Chromatographically purified as described by Frattali, V., and Steiner, R.: Biochem., 7, 521 (1968), modified by Worthington. A dialyzed, lyophilized powder. Purity >90% checked using SDS PAGE. Store at 2-8°C.