Learn More
HIMEDIA Soya Peptone Yeast Extract Agar, M935, Microbiology Culture Media, Dehydrated Culture Media, Selective Agar, Dermatophyte Isolation, Antibiotic Supplemented, Powder, 500 g
Catalog No. NC4013767
Encompass_Preferred
Soya Peptone Yeast Extract Agar is a selective medium for isolating dermatophytes, especially Trichophyton verrucosum, and other pathogenic fungi. Soya peptone, yeast extract and a high dextrose level promote fungal growth and pigmentation, while streptomycin sulphate and chloramphenicol suppress bacterial overgrowth, final pH 6.6. This combination makes it valuable in clinical and veterinary mycology for recovering slow-growing dermatophytes from skin, hair and nail samples. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013767 Supplier HIMEDIA Supplier No. M935500G
Soya Peptone Yeast Extract Agar is a selective medium for isolating dermatophytes, especially Trichophyton verrucosum, and other pathogenic fungi. Soya peptone, yeast extract and a high dextrose level promote fungal growth and pigmentation, while streptomycin sulphate and chloramphenicol suppress bacterial overgrowth, final pH 6.6. This combination makes it valuable in clinical and veterinary mycology for recovering slow-growing dermatophytes from skin, hair and nail samples. Dehydrated powder, store in a cool dry place. Packaged as 500 g.