Learn More
Medchemexpress LLC Soya phosphatidylcholines | 97281-47-5 | MFCD18633235 | 99.0% | 756.0874 | C43H82NO7P | 250MG
SDP
Catalog No. 5000234947
Encompass_Preferred
Phosphatidylcholines, soya (CAS 97281-47-5) is a soybean-derived phosphatidylcholine supplied as an oily material for use as a lipid component in liposome formulation and as a vehicle for administration in animal studies. The product is offered in research-scale pack sizes and is accompanied by supporting documentation for laboratory use.
Catalog No. 50-002-34947 Supplier Medchemexpress LLC Supplier No. HY125853250MG
Phosphatidylcholines, soya (CAS 97281-47-5) is a soybean-derived phosphatidylcholine supplied as an oily material for use as a lipid component in liposome formulation and as a vehicle for administration in animal studies. The product is offered in research-scale pack sizes and is accompanied by supporting documentation for laboratory use.