Learn More
Worthington Biochemical Corporation Trypsin Inhibitor, Soybean, 1g
SDP
Supplier:  Worthington Biochemical Corporation LS003587
Encompass
The soybean trypsin inhibitor was first crystallized by Kunitz in 1945 and is one of several such inhibitors found in soybeans. Its molecular weight is 21,500 ± 800 daltons and the optimum pH is 7.0. Soybean inhibitor inhibits trypsin mole-for-mole and to a lesser extent chymotrypsin. The soybean inhibitor is stable for 1-2 years at 2-8°C.
Catalog No. NC9065058