Learn More
Medchemexpress LLC Soyasapogenol A | 508-01-0 | 1 MG
SDP
Catalog No. 5000421662
Encompass_Preferred
Soyasapogenol A is a triterpenoid aglycone of soyasaponins. It exhibits various activities, including anti-inflammation, anti-cancer, hepatoprotection, and anti-HSV-1 effects. It can be utilized in research concerning tumors and immune inflammatory diseases.
  • Shows anti-inflammatory activity, as demonstrated by its ability to inhibit HMGB1-induced NO production in RAW264.7 cells.
  • Can inhibit the proliferation of Hep-G2 cells and induce cell cycle arrest and apoptosis.
  • Exerts ameliorative effects in a Concanavalin A-induced hepatitis model.
  • Possesses anti-HSV-1 activity.
  • Has a purity of 99.06%.
Catalog No. 50-004-21662 Supplier Medchemexpress LLC Supplier No. HYN60731MG
Add to Cart
Add to Cart
Soyasapogenol A is a triterpenoid aglycone of soyasaponins. It exhibits various activities, including anti-inflammation, anti-cancer, hepatoprotection, and anti-HSV-1 effects. It can be utilized in research concerning tumors and immune inflammatory diseases.
  • Shows anti-inflammatory activity, as demonstrated by its ability to inhibit HMGB1-induced NO production in RAW264.7 cells.
  • Can inhibit the proliferation of Hep-G2 cells and induce cell cycle arrest and apoptosis.
  • Exerts ameliorative effects in a Concanavalin A-induced hepatitis model.
  • Possesses anti-HSV-1 activity.
  • Has a purity of 99.06%.