Learn More
Medchemexpress LLC HY-N6074 5mg Medchemexpress, Soyasapogenol B CAS:595-15-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN60745MG
Encompass_Preferred
Medchemexpress, HY-N6074 5mg Soyasapogenol B CAS:595-15-3 Soyasapogenol B, an ingredient of soybean, exerts anti-proliferative, anti-metastatic activities. Soyasapogenol B triggers endoplasmic reticulum stress, which mediates apoptosis and autophagy in colorectal cancer [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3956