Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Soyasapogenol C | 595-14-2 | 5 MG
SDP

Catalog No. 5000418928
Encompass_Preferred

Soyasapogenol C is an oleanane-type triterpenoid that exhibits anti-HSV-1 activity with an IC50 of 18.9 μM. It is intended for research use only and is not sold to patients.

  • Oleanane-type triterpenoid
  • Exhibits anti-HSV-1 activity with an IC50 of 18.9 μM
  • No cytotoxicity at CC50 of 921 μM
  • Molecular weight: 440.70
  • Formula: C30H48O2
  • Appearance: White to off-white solid
  • For research use only
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 2.5 mg/mL (5.67 mM)

Catalog No. 50-004-18928 Supplier Medchemexpress LLC Supplier No. HYN81565MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Soyasapogenol C is an oleanane-type triterpenoid that exhibits anti-HSV-1 activity with an IC50 of 18.9 μM. It is intended for research use only and is not sold to patients.

  • Oleanane-type triterpenoid
  • Exhibits anti-HSV-1 activity with an IC50 of 18.9 μM
  • No cytotoxicity at CC50 of 921 μM
  • Molecular weight: 440.70
  • Formula: C30H48O2
  • Appearance: White to off-white solid
  • For research use only
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 2.5 mg/mL (5.67 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.