Learn More
Medchemexpress LLC Soyasaponin Aa | 117230-33-8 | 1 MG

Supplier: Medchemexpress LLC HYN30271MG
Soyasaponin Aa is a soyasaponin that exerts an anti-obesity effect in 3T3-L1 adipocytes through downregulation of peroxisome proliferator-activated receptor γ (PPARγ).
- Molecular formula: C64H100O31
- Molecular weight: 1365.46
- Target: PPAR
- Pathway: Cell Cycle/DNA Damage, Metabolic Enzyme/Protease, Vitamin D Related/Nuclear Receptor
- In vitro solubility: DMSO 100 mg/mL (73.24 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.