Learn More
Cayman Chemical Soyaspogenol B 10mg
SDP
Catalog No. 502942813
Encompass_Preferred
A triterpenoid with diverse biological activities; inhibits LPS-induced increases in the levels of NO in RAW 264.7 macrophages at 95.6 µM; decreases triglyceride levels, as well as increases glycerol and free fatty acids levels in, and decreases the levels of resistin secreted from, 3T3-L1 adipocytes at 12.5 µM; reduces plasma levels of ALT in a mouse model of concanavalin A-induced autoimmune hepatitis
Catalog No. 50-294-2813 Supplier Cayman Chemical Supplier No. 4027010 MG
A triterpenoid with diverse biological activities; inhibits LPS-induced increases in the levels of NO in RAW 264.7 macrophages at 95.6 µM; decreases triglyceride levels, as well as increases glycerol and free fatty acids levels in, and decreases the levels of resistin secreted from, 3T3-L1 adipocytes at 12.5 µM; reduces plasma levels of ALT in a mouse model of concanavalin A-induced autoimmune hepatitis