Learn More
Bioworld Glycine max (Soybean) Lectin (SBA) - FITC
Supplier:  Bioworld 217610241
Encompass
Technical Specifications:Ligand Source:Glycine maxMolecular Weight: 120 kDaCarbohydrate Specificity:GalNAc, GalNAcα1,3GalInhibiting/Eluting Sugar:N-AcetylgalactosamineCarbohydrate for Elution: 0.2MN-AcetylgalactosamineBlood Group Specificity: A>O>BConjugate: FITCForm: LiquidIsoelectric (pI): 5.8-6Storage: -20°CApplication note: Calcium and Manganese are required for binding.
Catalog No. 50-199-130