Learn More
MilliporeSigma Tryptic Soybean Broth (TSB), 50/pk
Catalog No. NC0273548
Encompass
A non-selective medium used to detect total heterotrophic microorganisms in water and other liquids.
Catalog No. NC0273548 Supplier MilliporeSigma Supplier No. MHA000T2B
A non-selective medium used to detect total heterotrophic microorganisms in water and other liquids.