Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Soybean oil | 8001-22-7 | Reagent grade | 100 MG
SDP

Catalog No. 5000344656
Encompass_Preferred

Soybean oil is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.

  • Liquid appearance
  • Colorless to light yellow
  • Store at 4°C, sealed, away from moisture
  • Soluble in DMSO at 100 mg/mL (requires ultrasonication)

Catalog No. 50-003-44656 Supplier Medchemexpress LLC Supplier No. HY10875010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Soybean oil is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.

  • Liquid appearance
  • Colorless to light yellow
  • Store at 4°C, sealed, away from moisture
  • Soluble in DMSO at 100 mg/mL (requires ultrasonication)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.