Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soybean Oil, MP Biomedicals™

Catalog No. MP210468705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210468705 5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210468705 Supplier MP Biomedicals, Inc Supplier No. 0210468705
Add to Cart
Add to Cart

Soybean Oil, Orange Oil

  • Soybean Oil may be used as a crude source for triglycerides and other lipids.
  • Soybean oil is a potential raw material in oxygenated fatty acid production.
  • Soybean oil (SBO) is a vegetable oil widely used as biodegradable lubricant.
  • It may be transesterified to biodiesel using CaO as a solid base catalyst.
  • It may also be transesterified to biodiesel using methanol and ethanol.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 001-22-7
MDL Number MFCD00132356
Synonym Soya oil

Specifications

Melting Point 15°F to 20°F(literature)
Color Orange
Density 0.917g/mL at 25°C , 0.92g/mL at 25°C (lit.)
Refractive Index n20/D 1.4743 (lit.)
Quantity 5 lb.
Solubility Information Miscible with alcohol, ether, petroleum ether; insoluble in water.
Physical Form Oil
RTECSNumber WG4862000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.