Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soybean Oil, USP, Spectrum™ Chemical
SDP

Catalog No. 18606966 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-966 20 L Poly Drum
18-606-967 200 L Drum
18-606-965 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-966 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO25520LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

SO255, 8001-22-7

Spectrum™ Chemical Soybean Oil, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8001-22-7

Specifications

CAS Max % 1
Packaging Poly Drum
Quantity 20 L
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.