Learn More
TARGETMOL CHEMICALS INC SP-13786 5MG
SDP
Catalog No. 502579110
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SP-13786 (UAMC-1110) is an inhibitor of fibroblast activation protein (FAP) (IC50 3.2 nM); also inhibits prolyl oligopeptidase (PREP) ( IC50 1.8 uM). purity: 99%
Catalog No. 50-257-9110 Supplier TARGETMOL CHEMICALS INC Supplier No. T78905MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SP-13786 (UAMC-1110) is an inhibitor of fibroblast activation protein (FAP) (IC50 3.2 nM); also inhibits prolyl oligopeptidase (PREP) ( IC50 1.8 uM). purity: 99%