Learn More
Medchemexpress LLC SP-141 | 1253491-42-7 | 98.9% | 324.38 | 50 MG
SDP
Catalog No. 5000343134
Encompass_Preferred
SP-141 is a specific inhibitor of MDM2 that promotes MDM2 auto-ubiquitination and degradation. It is used for research on pancreatic and breast cancer cells, inhibiting human pancreatic cancer cell growth in a p53-independent manner and showing selective cytotoxicity for cancer cells.
  • Promotes MDM2 auto-ubiquitination and degradation.
  • Inhibits human pancreatic cancer cell growth.
  • Shows selective cytotoxicity for cancer cells.
  • Suppresses pancreatic tumor growth in xenograft and orthotopic mouse models.
Catalog No. 50-003-43134 Supplier Medchemexpress LLC Supplier No. HY11018210MM/1ML
SP-141 is a specific inhibitor of MDM2 that promotes MDM2 auto-ubiquitination and degradation. It is used for research on pancreatic and breast cancer cells, inhibiting human pancreatic cancer cell growth in a p53-independent manner and showing selective cytotoxicity for cancer cells.
  • Promotes MDM2 auto-ubiquitination and degradation.
  • Inhibits human pancreatic cancer cell growth.
  • Shows selective cytotoxicity for cancer cells.
  • Suppresses pancreatic tumor growth in xenograft and orthotopic mouse models.