Learn More
Abcam SP-2509, LSD1 inhibitor, 5MG
Catalog No. 502672391
Encompass_Preferred
MW 437.9 Da, Purity >98%. Potent, selective (IC₅₀ = 13 nM) and reversible inhibitor of the histone demethylase LSD1 (KDM1A). No activity against MAO-A, MAO-B, lactate dehydrogenase and glucose oxidase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2391 Supplier Abcam Supplier No. AB2544595MG
MW 437.9 Da, Purity >98%. Potent, selective (IC₅₀ = 13 nM) and reversible inhibitor of the histone demethylase LSD1 (KDM1A). No activity against MAO-A, MAO-B, lactate dehydrogenase and glucose oxidase.
The product is subject to the following: Abcam Restricted Use Statement