Learn More
Cayman Chemical Sp-5,6-dichloro-cBIMPS (sodium salt), Purity: ≥99%, CAS: 142439-96-1, 5 mg
SDP
Catalog No. 502967935
Encompass_Preferred
A potent activator of PKA (Ki = 30 nM) that is selective for PKA over PKG (Ki = 10 µM); resistant to degradation by cyclic nucleotide phosphodiesterases, displays good cell permeability, and prevents the aggregation of platelets stimulated with thrombin
Catalog No. 50-296-7935 Supplier Cayman Chemical Supplier No. 188225 MG
A potent activator of PKA (Ki = 30 nM) that is selective for PKA over PKG (Ki = 10 µM); resistant to degradation by cyclic nucleotide phosphodiesterases, displays good cell permeability, and prevents the aggregation of platelets stimulated with thrombin