Learn More
Apexbio Technology LLC SP 600125 129-56-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061426
Encompass_Preferred
SP 600125 (CAS 129-56-6) is a selective reversible ATP-competitive inhibitor of c-Jun N-terminal kinase (JNK) with reported IC50 values of 40 nM for JNK1 and JNK2 and 90 nM for JNK3 Initially identified via GST-c-Jun substrate and recombinant human JNK2 screening assays SP 600125 demonstrates marked specificity being approximately 300-fold more selective toward JNK isoforms compared to ERK1 and p38-2 kinases In cellular assays SP 600125 effectively suppresses JNK-mediated phosphorylation of c-Jun and attenuates expression of IL-2 IFN- and TNF- It is widely utilized in inflammation and signal transduction research
Catalog No. 50-000-61426 Supplier Apexbio Technology LLC Supplier No. A46045.1
SP 600125 (CAS 129-56-6) is a selective reversible ATP-competitive inhibitor of c-Jun N-terminal kinase (JNK) with reported IC50 values of 40 nM for JNK1 and JNK2 and 90 nM for JNK3 Initially identified via GST-c-Jun substrate and recombinant human JNK2 screening assays SP 600125 demonstrates marked specificity being approximately 300-fold more selective toward JNK isoforms compared to ERK1 and p38-2 kinases In cellular assays SP 600125 effectively suppresses JNK-mediated phosphorylation of c-Jun and attenuates expression of IL-2 IFN- and TNF- It is widely utilized in inflammation and signal transduction research