Learn More
V&P Scientific Inc SpinVessel, 6L
Catalog No. NC2068067
Encompass
SpinVessel, 6L, Round Bottom, Disposable, Sterile, RNase, DNase and DNA Free, Nonpyrogenic, Virgin Polypropylene, with Pulsed Radial Flow Side Fins and Projections, Individually Packaged and Sealed, US Patent #11,623,188, European Patent #3887049
Catalog No. NC2068067 Supplier V&P Scientific Inc Supplier No. VP 830SV6L
SpinVessel, 6L, Round Bottom, Disposable, Sterile, RNase, DNase and DNA Free, Nonpyrogenic, Virgin Polypropylene, with Pulsed Radial Flow Side Fins and Projections, Individually Packaged and Sealed, US Patent #11,623,188, European Patent #3887049