Learn More
Enzo Life Sciences Sp-8-Br-cGMPS (500µg). CAS: 153660-03-8
SDP
Supplier:  Enzo Life Sciences BMLCN2170500
Encompass_Preferred
Cell-permeable, phosphodiesterase-resistant cGMP agonist. This analog activates PKA and PKG with similar potency. Much more lipophilic and membrane permeant as compared to cyclic GMP (ALX-480-059) or Sp-cGMPS (ALX-480-073) Purity: ≥98% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0456